missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Replacement Needle for CTC PAL RTC LC Syringes

Reduce costs. Replace the needle not the syringe. For PAL RTC instruments.

Brand:  Trajan Scientific and Medical™ 039280

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 13139035

  • 328.00 DKK / Pack of 2
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Increased application specific flexibility
  • Reduced overall costs by replacing the needle rather than the entire syringe
Specifications

Specifications

0.41 mm
57 mm
2/Pk.
CTC RTC and Thermo RSH Syringes (1 to 2.5mL )
0.72 mm
Gastight
LC
Fixed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers