Learn More
WTW™ Replacement Flow Through Vessel
Perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine to conductivity of solutions in closed circuits.
Brand: WTW™ 302760
Additional Details : Weight : 1.00000kg
Description
- Overpressure must not exceed 0.5 bar
- For conductivity measuring cells for applications to prevent air from entering
- Manufactured from stainless steel, for LR 325/001
Specifications
Replacement Flow Through Vessel | |
Preventing air entry, closed circuit measurements | |
Stainless Steel | |
LR 325/001 |