Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Brand:  Corning™ 6621
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15853895
  • 384.00 DKK / Pack of 100
Estimated Shipment
11-03-2019
Add to basket
Description
Description
  • Polypropylene tips for use with the Corning™ Step-R™ Repeating Pipettor
  • Available in eight sizes from 0.05mL to 50mL
  • Tips use the direct displacement method, dedicated for use with viscous, infectious and volatile liquids
  • Reliable and repeatable results through automated, individual tests of every single tip
  • Certification of every batch is available on request
Specifications
Specifications
Yes
Polypropylene
0.5mL
100/PK
Non-sterile
Dispensing of viscous and volatile liquids
ISO8655
Videos