Få mere at vide
Tocris™ Reparixin L-lysine salt
Beskrivelse
Reparixin L-lysine salt is a potent and noncompetitive CXCR1 and CXCR2 allosteric antagonist (IC50 = 1 nM for inhibition of CXCL8-induced human polymorphonuclear cell migration). Also inhibits migration of rodent neutrophils induced by CXCL1, CXCL2, CXCL8 and CINC-1. Inhibits vascular permeability and neutrophil recruitment in in vivo models of mild and severe ischemia/reperfusion injury. Also selectively depletes cancer stem cells in human breast cancer cell lines and xenograft models.
Tekniske data
Tekniske data
| CAS | 266359-93-7 |
| Synonym | Repertaxin L-lysine salt |
| Mængde | 10 mg |
| Opløselighed | Soluble to 50 mM in DMSO and to 5 mM in water with gentle warming |
| Molekylær formel | C14H21NO3S.C6H14N2O2 |
| Formel vægt | 429.58 |
| Produkttype | Antagonist |
| Renhed | 98% |