missing translation for 'onlineSavingsMsg'
Få mere at vide

Renal Cell Carcinoma Marker (gp200) Ab-1 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Renal Cell Carcinoma Marker (gp200) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-409-P1ABX

 Se flere versioner af dette produkt

Produktkode 12631367

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Of other normal tissues examined, the gp200 is also localized along the luminal surfaces of breast lobules and ducts, the luminal surface of the epididymal tubular epithelium, within the cytoplasm of parathyroid parenchymal cells, and focally within the colloid of thyroid follicles. Thirty-one other normal tissues do not express similar or cross-reacting antigens. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas.

Host Species: Mouse

Clone: PN-15

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Carbohydrate domain

Immunogen: Microsomal fraction of human renal cortical tissue homogenate

Molecular Weight: ∽200kDa

Positive Control: Normal kidney or renal cell CA

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Reduced and nonreduced)
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Renal Cell Carcinoma Marker (gp200) Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Cancer and Tumor Biology
Human, Rat
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
PN-15
Unconjugated
Microsomal fraction of human renal cortical tissue homogenate
IVD
Primary
Concentrated
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.