Learn More
Trajan Scientific and Medical™ Removable Needle for Thermo Scientific RSH Autosampler
Replacement needles for use with Thermo Scientific RSH Autosampler
Brand:  Trajan Scientific and Medical™ 037873
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13109185
  • 396.00 DKK / Pack of 2
Estimated Shipment: 01-07-2024
to see stock.
Description
Description
  • Replacement needle for use with Thermo Scientific RSH Autosampler
  • Removable needle
  • Cone Tip
Specifications
Specifications
23 G
85 mm
10 μL
Thermo RSH Autosampler
0.63 mm
Removable Needle
Cone