Få mere at vide
Tocris Bioscience™ Remodelin
N-acetyltransferase 10 (NAT10) inhibitor
Brand: Tocris Bioscience™ 5193/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Remodelin is a N-acetyltransferase 10 (NAT10) inhibitor. Improves nuclear shape in HGPS (Hutchinson-Gilford progeria syndrome) cells and lamin A/C depleted cells.Tekniske data
| 4-[2-(2-Cyclopentylidenehydrazinyl)-4-thiazolyl]benzonitrile | |
| 949912-58-7 | |
| 10 mg |