missing translation for 'onlineSavingsMsg'
Få mere at vide

Remodelin hydrobromide, MedChemExpress

Artikelnummer. 16499677
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16499677 5 mg
16467148 10 mg
16433177 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16499677 Leverandør Medchem Express Leverandørnr. HY16706A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Remodelin, a specific inhibitor of N-acetyltransferase NAT10, can ameliorate Hutchinson-Gilford Progeria Syndrom (HGPS) cellular phenotypes. Remodelin acts in a progerin- and FTI-independent pathway, by targeting and inhibiting NAT10. NAT10 is a protein with histone acetylation activity and primarily identified to be involved in regulation of telomerase activity.

Remodelin, a specific inhibitor of N-acetyltransferase NAT10, can ameliorate Hutchinson-Gilford Progeria Syndrom (HGPS) cellular phenotypes. Remodelin acts in a progerin- and FTI-independent pathway, by targeting and inhibiting NAT10. NAT10 is a protein with histone acetylation activity and primarily identified to be involved in regulation of telomerase activity.

Tekniske data

Kemisk navn eller materiale Remodelin hydrobromide
Fysisk form Solid
CAS 1622921-15-6
Farve Yellow
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C15H15BrN4S
Til brug med (applikation) Cancer-programmed cell death
Opløselighedsinformation DMSO : ≥ 44 mg/mL (121.12 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL N#CC1=CC=C(C2=CSC(N/N=C3CCCC/3)=N2)C=C1.[H]Br
Molekylvægt (g/mol) 363.28
Formel vægt 363.28
Procent renhed 99.16%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.