Learn More
Thermo Scientific™ Remel™ Gram Decolorizer
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
305.00 DKK - 1605.00 DKK
Product Code | Brand | Type | Quantity | Certifications/Compliance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Certifications/Compliance | Price | Quantity & Availability | |||
11922942
|
Thermo Scientific™
R40054 |
Decolorizer | Each | CE Marked |
305.00 DKK
250mL |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11972942
|
Thermo Scientific™
R40055 |
pYR Reagent Dropper | 5/Pk. | CE Marked |
1125.00 DKK
Pack of 5 |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11924092
|
Thermo Scientific™
R40075 |
Decolorizer | Each | CE Marked |
1605.00 DKK
4 L |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Thermo Scientific™ Remel Gram Decolorizer is a decolorizing reagent used in Gram stain procedures.
- For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
- Selected Gram stains are CE marked
Specifications
CE Marked |