Få mere at vide
rel-(4R,5R)-5-(4-Chlorophenyl)-4-methyl-2-thiazolidinone, TRC
Artikelnummer. 17931465
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17931465 10 mg
17993265 100 mg
17983655 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17931465 Leverandør Toronto Research Chemicals Leverandørnr. TRCC54695010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 78587-59-4
Molekylær formel C10 H10 Cl N O S
Molekylvægt (g/mol) 227.71
Synonym 2-Thiazolidinone, 5-(4-chlorophenyl)-4-methyl-, trans-, rel-(4R,5R)-5-(4-Chlorophenyl)-4-methyl-2-thiazolidinone, trans-5-(4-Chlorophenyl)-4-methyl-2-thiazolidinone, Hexythiazox metabolite PT-1-3
IUPAC navn (4S,5S)-5-(4-chlorophenyl)-4-methyl-1,3-thiazolidin-2-one
SMIL C[C@@H]1NC(=O)S[C@H]1c2ccc(Cl)cc2
Tekniske data
Kemisk navn eller materiale rel-(4R,5R)-5-(4-Chlorophenyl)-4-methyl-2-thiazolidinone
Mængde 10 mg
InChI formel InChI=1S/C10H10ClNOS/c1-6-9(14-10(13)12-6)7-2-4-8(11)5-3-7/h2-6,9H,1H3,(H,12,13)/t6-,9+/m0/s1
Formel vægt 227.0172