Få mere at vide
rel-(1R,3S)-cyclopentane-1,3-dicarboxylic Acid, TRC
Artikelnummer. 30422019
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422019 250 mg
30418067 500 mg
30419378 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30422019 Leverandør Toronto Research Chemicals Leverandørnr. TRCR160711250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 876-05-1
Molekylær formel C7H10O4
Molekylvægt (g/mol) 158.15
Synonym rel-(1R,3S)-1,3-Cyclopentanedicarboxylic Acid, 1,3-cis-Cyclopentanedicarboxylic Acid, NSC 42324, NSC 45402, cis-1,3-Cyclopentanedicarboxylic Acid, cis-1,3-Cyclopentanedioic Acid
IUPAC navn (1R,3S)-cyclopentane-1,3-dicarboxylic acid
SMIL OC(=O)[C@@H]1CC[C@@H](C1)C(=O)O
Tekniske data
Kemisk navn eller materiale rel-(1R,3S)-cyclopentane-1,3-dicarboxylic Acid
Mængde 250 mg
InChI formel InChI=1S/C7H10O4/c8-6(9)4-1-2-5(3-4)7(10)11/h4-5H,1-3H2,(H,8,9)(H,10,11)/t4-,5+
Formel vægt 158.0579