Learn More
rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate, 95%, BLD Pharm™
Product Code. 31447335
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31447335 100 g
31447173 500 g
31437265 10 g
31445737 1 kg
31438226 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31447335 Supplier BLD Pharm Supplier No. BD135009100g
 Restricted Item
Forventet forsendelse: 25-11-2026
Stabilized 150 to 200 ppm MEHQ
Specifications
CAS 7534-94-3
Molecular Formula C14H22O2
MDL Number MFCD00081070
Quantity 100 g
SMILES CC(C(O[C@H]1[C@](C2(C)C)(C)CC[C@]2([H])C1)=O)=C
Molecular Weight (g/mol) 222.32
Percent Purity 95%
Chemical Name or Material rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate