Learn More
BINDER™ Heating Output, Reinforced
Heating output, reinforced - 400 V option model, for faster heating up time and improved recovery time; max. 300°C
Brand:  BINDER™ 8012-1061
Product Code. 16494049
  • 8030.00 DKK / Each
Only null left
Estimated Shipment: 28-08-2025
Description
  • 400 V option model
  • Heats up faster to temperatures up to 300°C
  • Improves recovery time
Specifications
Specifications
Heating output, reinforced - 400 V option model, for faster heating up time and improved recovery time; max. 300°C
300°C (Max.)
For Faster Heating up Time and Improved Recovery Time
BINDER Chambers FD 720 Drying and Heating Chambers with Forced Convection
Heating Output
400 V
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Heating Output, Reinforced > Reinforced Heating Output; For Use With (Equipment): BINDER™ FD 720 Drying and Heating Chambers with Forced Convection