missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg4 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99734R
Additional Details : Weight : 0.00970kg
Description
Reg4 Polyclonal specifically detects Reg4 in Human samples. It is validated for ELISA.Specifications
Reg4 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 550 | |
Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein | |
Produced in rabbits immunized with purified, recombinant Reg4 (NP_001152824.1; Met 1-Pro 158). | |
0.1 mL | |
Immunology | |
83998 | |
Store at 4°C in the dark. | |
IgG |