Få mere at vide
Techne™ Refrigerated Bath
Model-:
RB-12A
RB-5A
Kapacitet (metrisk):
7 l
12 l
Pakningsstørrelse:
Hver
Beskrivelse
- Built-in refrigeration for efficient cooling
- Robust stainless steel construction makes for easy cleaning
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 7 l |
| Kølekapacitet | 20°C at 145W, 0°C at 145W, -10° C at 110W |
| Temperaturområde | -20°C to +100°C |
| Funktioner | Circulating bath with integrated refrigeration unit, Temperature range from -35°C to +100°C, Robust stainless-steel construction, Three models of refrigerated bath available: 7, 12, 22 liter,3 year warranty. |
| Model- | RB-5A |
| garanti | 3 Years |
| Spænding | 230 V |
| Frekvens | 50/60 Hz |