Learn More
Lenz Laborglasintrumente Reduction Adapters with Ground Joints
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01502971
Additional Details : Weight : 0.40000kg
Description
- Acc. to DIN 12257 and in addition to DIN standard
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifications
NS 71/51 | |
Borosilicate Glass (3.3) | |
Socket NS 29/32 |