Learn More
BOLA Reducing Unions
Reducing Unions PTFE, glass-fibre reinforced PTFE nut
Brand:  BOLA D526-36
 View more versions of this product
Product Code. 10609601
  • 662.00 DKK / Each
Only null left
Estimated Shipment: 31-07-2025
Description
Straight tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE. For connecting tubing or tube with different outer diameters. Universal chemical resistance, the product is only exposed to PTFE.
Specifications
Specifications
FDA
16 mm
White
Autoclavable
6 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BOLA Reducing Unions > 16 mm