missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA Reducing Unions
Reducing Unions PTFE, glass-fibre reinforced PTFE nut
Brand: BOLA D526-28
This item is not returnable.
View return policy
Description
Straight tube fitting made of PTFE with nuts made of glass-fibre reinforced PTFE. For connecting tubing or tube with different outer diameters. Universal chemical resistance, the product is only exposed to PTFE.
Specifications
FDA | |
14 mm | |
White |
Autoclavable | |
4 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BOLA Reducing Unions > 14 mm
Spot an opportunity for improvement?Share a Content Correction