Få mere at vide
Reduced Haloperidol, TRC
Artikelnummer. 30137055
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137055 50 mg
30003283 25 mg
30005528 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137055 Leverandør Toronto Research Chemicals Leverandørnr. TRCR14240050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34104-67-1
Molekylær formel C21 H25 Cl F N O2
Molekylvægt (g/mol) 377.88
IUPAC navn 4-(4-chlorophenyl)-1-[4-(4-fluorophenyl)-4-hydroxybutyl]piperidin-4-ol
SMIL OC(CCCN1CCC(O)(CC1)c2ccc(Cl)cc2)c3ccc(F)cc3
Tekniske data
Kemisk navn eller materiale Reduced Haloperidol
Mængde 50 mg
InChI formel InChI=1S/C21H25ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,20,25-26H,1-2,11-15H2
Formel vægt 377.1558