Få mere at vide
Red fluorescent protein Mouse anti-Recombinant Protein, Unconjugated, Clone: 1E3C5, Proteintech
Artikelnummer. 16819975 Shop alle Proteintech produkter
Skift visning
Mængde:
150 μL
Artikelnummer. Mængde
16819975 150 μL
1 options
Artikelnummer. 16819975 Leverandør Proteintech Leverandørnr. 673781IG150UL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Mouse Monoclonal Antibody
The Red fluorescent protein antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of entacmaea quadricolor Red fluorescent protein. This antibody recognizes recombinant protein antigen. The Red fluorescent protein antibody has been validated for the following applications: WB, ELISA analysis.
Tekniske data
Antigen Red fluorescent protein
Applikationer Western Blot, ELISA
Klassifikation Monoclonal
Klon 1E3C5
Koncentration 1000 μg/mL
Konjugeret Unconjugated
Værtsarter Mouse
Mængde 150 μL
Primær eller sekundær Primary
Isotype IgG2b