missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zebrafish Metallothionein-2 Protein

A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) Metallothionein-2, was expressed in the hosts and tags indicated.

2615.00 DKK - 23570.00 DKK

Specifications

Accession Number NP_001124525.2
Gene ID (Entrez) 100174951
Name Metallothionein-2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16038791
View Documents
enQuireBio™
QP9826-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16078791
View Documents
enQuireBio™
QP9826-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028791
View Documents
enQuireBio™
QP9826-ye-100ug
100 ÎĽg
7930.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16058791
View Documents
enQuireBio™
QP9826-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16068791
View Documents
enQuireBio™
QP9826-ye-500ug
500 ÎĽg
19165.00 DKK
500µg
Please to purchase this item. Need a web account? Register with us today!
16048791
View Documents
enQuireBio™
QP9826-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_001124525.2
Metallothionein-2 Protein
Not determined.
Full Length
Zebrafish
His
Tris-based buffer, 50% glycerol
100174951
Research Use Only
mt2
Recombinant Protein
Yeast
MDPCECAKTG TCNCGATCKC TNCQCTTCKK SCCSCCPSGC SKCASGCVCK GNSCGSSCCQ
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Zebrafish Metallothionein-2 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.