missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Xenopus Protein Wnt-8 Protein
A DNA sequence encoding the Xenopus laevis (African clawed frog) Protein Wnt-8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9601-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001081637.1 | |
Protein Wnt-8 Protein | |
Research Use Only | |
wnt8 | |
Recombinant Protein | |
Yeast | |
AWSVNNFLMT GPKAYLTYSA SVAVGAQNGI EECKYQFAWE RWNCPESTLQ LATHNGLRSA TRETSFVHAI SSAGVMYTLT RNCSMGDFDN CGCDDSRNGR IGGRGWVWGG CSDNAEFGER ISKLFVDGLE TGQDARALMN LHNNEAGRLA VKETMKRTCK CHGISGSCSI QTCWLQLAEF RDIGNHLKIK HDQALKLEMD KRKMRSGNSA DNRGAIADAF SSVAGSELIF LEDSPDYCLK NISLGLQGTE GRECLQSGKN LSQWERRSCK RLCTDCGLRV EEKKTEIISS CNCKFHWCCT VKCEQCKQVV IKHFCARRER DSNMLNTKRK NRGHRR | |
Greater than 90% as determined by SDS-PAGE. |
397970 | |
10 μg | |
Not determined. | |
Full Length | |
Xenopus | |
His | |
Tris-based buffer, 50% glycerol |