missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Xenopus Protein Wnt-8 Protein

A DNA sequence encoding the Xenopus laevis (African clawed frog) Protein Wnt-8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9601-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018511

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001081637.1
Protein Wnt-8 Protein
Research Use Only
wnt8
Recombinant Protein
Yeast
AWSVNNFLMT GPKAYLTYSA SVAVGAQNGI EECKYQFAWE RWNCPESTLQ LATHNGLRSA TRETSFVHAI SSAGVMYTLT RNCSMGDFDN CGCDDSRNGR IGGRGWVWGG CSDNAEFGER ISKLFVDGLE TGQDARALMN LHNNEAGRLA VKETMKRTCK CHGISGSCSI QTCWLQLAEF RDIGNHLKIK HDQALKLEMD KRKMRSGNSA DNRGAIADAF SSVAGSELIF LEDSPDYCLK NISLGLQGTE GRECLQSGKN LSQWERRSCK RLCTDCGLRV EEKKTEIISS CNCKFHWCCT VKCEQCKQVV IKHFCARRER DSNMLNTKRK NRGHRR
Greater than 90% as determined by SDS-PAGE.
397970
200 μg
Not determined.
Full Length
Xenopus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers