missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL10 / Interleukin-10 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9777-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001009327.1 | |
IL10 / Interleukin-10 Protein | |
Research Use Only | |
IL10 | |
Recombinant Protein | |
Yeast | |
SRDASTLSDS SCTHFPASLP HMLRDVRAAF GKVKTFFQMK DQLNSMLLTQ SLLDDFKGYL GCQALSEMIQ FYLEEVMPQA ENHGPDIKEH VNSLGEKLKT LRLRLRRCHR FLPCENKSKA VEQVKRVFNM LQERGVYKAM SEFDIFINYI ESYMTTKM | |
Greater than 90% as determined by SDS-PAGE. |
443342 | |
50 μg | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |