Få mere at vide
enQuireBio™ Recombinant S. cerevisiae DNA repair and recombination protein RAD52 Protein
Artikelnummer. p-7154070
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16078470
Brand: enQuireBio™ QP7366ec10ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) DNA repair and recombination protein RAD52, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_013680.2
Gen-id (Entrez) 854976
Navn S. cerevisiae DNA repair and recombination protein RAD52 Protein
Mængde 10 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol RAD52
Protein længde Partial (see sequence information for more details).
Produkttype Recombinant Protein
Krydsreaktivitet S. cerevisiae
Arter E. coli
Protein tag His-SUMO
Sekvens IFGYNGWSTE VKSVVIDFLD ERQGKFSIGC TAIVRVTLTS GTYREDIGYG TVENERRKPA AFERAKKSAV TDALKRSLRG FGNALGNCLY DKDFLAKIDK VKFDPPDFDE NNLFRPTDEI SESSRTNTLH ENQEQQQYPN KRRQLTKVTN TNPDSTKNLV KIENTVSRGT PMMAAPAEAN SKNSSNKDTD LKSLDASKQD QDDLLDDSLM FSDDFQDDDL INM
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.