missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Alanine / arginine aminopeptidase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Alanine/arginine aminopeptidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7127-100ug
This item is not returnable.
View return policy
Specifications
NP_011913.1 | |
S. cerevisiae Alanine / arginine aminopeptidase Protein | |
Research Use Only | |
AAP1 | |
Recombinant Protein | |
E. coli | |
MSREVLPNNV TPLHYDITLE PNFRAFTFEG SLKIDLQIND HSINSVQINY LEIDFHSARI EGVNAIEVNK NENQQKATLV FPNGTFENLG PSAKLEIIFS GILNDQMAGF YRAKYTDKVT GETKYMATTQ MEATDARRAF PCFDEPNLKA TFAVTLVSES FLTHLSNMDV RNETIKEGKK YTTFNTTPKM STYLVAFIVA DLRYVESNNF RIPVRVYSTP GDEKFGQFAA NLAARTLRFF EDTFNIEYPL PKMDMVAVHE FSAGAMENWG LVTYRVIDLL LDIENSSLDR IQRVAEVIQH ELAHQWFGNL VTMDWWEGLW LNEGFATWMS WYSCNKFQPE WKVWEQYVTD NLQRALNLDS LRSSHPIEVP VNNADEINQI FDAISYSKG | |
Greater than 80% as determined by SDS-PAGE. |
856443 | |
100 ÎĽg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. cerevisiae Alanine / arginine aminopeptidase Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction