Få mere at vide
enQuireBio™ Recombinant S. cerevisiae Alanine / arginine aminopeptidase Protein
Artikelnummer. 16027460
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16027460
Brand: enQuireBio™ QP712750ug
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Alanine/arginine aminopeptidase, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_011913.1
Gen-id (Entrez) 856443
Navn S. cerevisiae Alanine / arginine aminopeptidase Protein
Mængde 50 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol AAP1
Protein længde Partial (see sequence information for more details).
Produkttype Recombinant Protein
Krydsreaktivitet S. cerevisiae
Arter E. coli
Protein tag His-SUMO
Sekvens MSREVLPNNV TPLHYDITLE PNFRAFTFEG SLKIDLQIND HSINSVQINY LEIDFHSARI EGVNAIEVNK NENQQKATLV FPNGTFENLG PSAKLEIIFS GILNDQMAGF YRAKYTDKVT GETKYMATTQ MEATDARRAF PCFDEPNLKA TFAVTLVSES FLTHLSNMDV RNETIKEGKK YTTFNTTPKM STYLVAFIVA DLRYVESNNF RIPVRVYSTP GDEKFGQFAA NLAARTLRFF EDTFNIEYPL PKMDMVAVHE FSAGAMENWG LVTYRVIDLL LDIENSSLDR IQRVAEVIQH ELAHQWFGNL VTMDWWEGLW LNEGFATWMS WYSCNKFQPE WKVWEQYVTD NLQRALNLDS LRSSHPIEVP VNNADEINQI FDAISYSKG
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 80% as determined by SDS-PAGE.