missing translation for 'onlineSavingsMsg'
Få mere at vide

enQuireBio™ Recombinant S. cerevisiae Alanine / arginine aminopeptidase Protein

Artikelnummer. 16007460
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16007460

Brand: enQuireBio™ QP7127200ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Alanine/arginine aminopeptidase, was expressed in the hosts and tags indicated.

Tekniske data

Adgangsnummer NP_011913.1
Gen-id (Entrez) 856443
Navn S. cerevisiae Alanine / arginine aminopeptidase Protein
Mængde 200 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol AAP1
Protein længde Partial (see sequence information for more details).
Produkttype Recombinant Protein
Krydsreaktivitet S. cerevisiae
Arter E. coli
Protein tag His-SUMO
Sekvens MSREVLPNNV TPLHYDITLE PNFRAFTFEG SLKIDLQIND HSINSVQINY LEIDFHSARI EGVNAIEVNK NENQQKATLV FPNGTFENLG PSAKLEIIFS GILNDQMAGF YRAKYTDKVT GETKYMATTQ MEATDARRAF PCFDEPNLKA TFAVTLVSES FLTHLSNMDV RNETIKEGKK YTTFNTTPKM STYLVAFIVA DLRYVESNNF RIPVRVYSTP GDEKFGQFAA NLAARTLRFF EDTFNIEYPL PKMDMVAVHE FSAGAMENWG LVTYRVIDLL LDIENSSLDR IQRVAEVIQH ELAHQWFGNL VTMDWWEGLW LNEGFATWMS WYSCNKFQPE WKVWEQYVTD NLQRALNLDS LRSSHPIEVP VNNADEINQI FDAISYSKG
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 80% as determined by SDS-PAGE.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.