Learn More
enQuireBio™ Recombinant S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) 2-deoxyglucose-6-phosphate phosphatase 1, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP9588-ye-100ug
 View more versions of this product
Product Code. 16028501
  • 7930.00 DKK / 100µg
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
NP_011910.1
S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein
Research Use Only
DOG1
Recombinant Protein
Yeast
MAEFSADLCL FDLDGTIVST TVAAEKAWTK LCYEYGVDPS ELFKHSHGAR TQEVLRRFFP KLDDTDNKGV LALEKDIAHS YLDTVSLIPG AENLLLSLDV DTETQKKLPE RKWAIVTSGS PYLAFSWFET ILKNVGKPKV FITGFDVKNG KPDPEGYSRA RDLLRQDLQL TGKQDLKYVV FEDAPVGIKA GKAMGAITVG ITSSYDKSVL FDAGADYVVC DLTQVSVVKN NENGIVIQVN NPLTRA
Greater than 85.6% as determined by SDS-PAGE.
856440
100 ÎĽg
Not determined.
Full. Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein > 100ÎĽg