missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) 2-deoxyglucose-6-phosphate phosphatase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9588-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068501

  • 17510.00 DKK / 500µg
Estimated Shipment: 03-08-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_011910.1
S. cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 Protein
500 μg
Not determined.
Full. Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
856440
Greater than 85.6% as determined by SDS-PAGE.
Research Use Only
DOG1
Recombinant Protein
Yeast
MAEFSADLCL FDLDGTIVST TVAAEKAWTK LCYEYGVDPS ELFKHSHGAR TQEVLRRFFP KLDDTDNKGV LALEKDIAHS YLDTVSLIPG AENLLLSLDV DTETQKKLPE RKWAIVTSGS PYLAFSWFET ILKNVGKPKV FITGFDVKNG KPDPEGYSRA RDLLRQDLQL TGKQDLKYVV FEDAPVGIKA GKAMGAITVG ITSSYDKSVL FDAGADYVVC DLTQVSVVKN NENGIVIQVN NPLTRA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers