missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Transposase for transposon Tn552 Protein
A DNA sequence encoding the Staphylococcus aureus Transposase for transposon Tn552, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7085-10ug
This item is not returnable.
View return policy
Specifications
WP_000790705.1 | |
S. aureus Transposase for transposon Tn552 Protein | |
Research Use Only | |
Tn552 | |
Recombinant Protein | |
E. coli | |
MKNKEKYLTN FSEAKRKEAT QKYNIIKPFI LGKQSLSSIS KSKGIALSTL YRWNKLYKEQ GLTGLIHNTR VDKGEHKLKQ NIIDEIKRLA LKNKRNSIAT IHRKIANYCI ENNFYKPSYK QVYSIIKAMP KSVIDFSHQG EKYYQNKYDL IQIRESSRPN EIWQADHTLL DIYILDQKGN INRPWLTIIM DDYSRAIAGY FISFDAPNAQ NTALTLHQAI WNKNNTNWPV CGIPEKFYTD HGSDFTSHHM EQVAIDLKIN LMFSKVGVPR GRGKIERFFQ TVNQTFLEQL PGYINNNDTS SDLIDFQNFE EKLRYFLIED YNQKEHSAIQ STPINRWNSN HFFPNMPSSL EQLDLLLLEI PKSRKIHSDG IHFQGFRYSN TNLTAYVGEY VLIRYNPNDM AEIRVFYRDE FLCTAISPDL ADYSIDIKEI QHARSQRRKH LKQNIASPST TDLIKEEKSY GYSPQETTKN VKKLKRYRND | |
Greater than 90% as determined by SDS-PAGE. |
13875815 | |
10 ÎĽg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. aureus Transposase for transposon Tn552 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction