missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Transposase for transposon Tn552 Protein

A DNA sequence encoding the Staphylococcus aureus Transposase for transposon Tn552, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7085-100ug

 View more versions of this product

Product Code. 16057330

  • 7745.00 DKK / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

WP_000790705.1
S. aureus Transposase for transposon Tn552 Protein
Research Use Only
Tn552
Recombinant Protein
E. coli
MKNKEKYLTN FSEAKRKEAT QKYNIIKPFI LGKQSLSSIS KSKGIALSTL YRWNKLYKEQ GLTGLIHNTR VDKGEHKLKQ NIIDEIKRLA LKNKRNSIAT IHRKIANYCI ENNFYKPSYK QVYSIIKAMP KSVIDFSHQG EKYYQNKYDL IQIRESSRPN EIWQADHTLL DIYILDQKGN INRPWLTIIM DDYSRAIAGY FISFDAPNAQ NTALTLHQAI WNKNNTNWPV CGIPEKFYTD HGSDFTSHHM EQVAIDLKIN LMFSKVGVPR GRGKIERFFQ TVNQTFLEQL PGYINNNDTS SDLIDFQNFE EKLRYFLIED YNQKEHSAIQ STPINRWNSN HFFPNMPSSL EQLDLLLLEI PKSRKIHSDG IHFQGFRYSN TNLTAYVGEY VLIRYNPNDM AEIRVFYRDE FLCTAISPDL ADYSIDIKEI QHARSQRRKH LKQNIASPST TDLIKEEKSY GYSPQETTKN VKKLKRYRND
Greater than 90% as determined by SDS-PAGE.
13875815
100 ÎĽg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant S. aureus Transposase for transposon Tn552 Protein > 100ÎĽg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.