missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Peptide deformylase Protein
A DNA sequence encoding the Staphylococcus aureus Peptide deformylase, was expressed in the hosts and tags indicated.
2615.00 DKK - 29485.00 DKK
Specifications
Accession Number | WP_000957037.1 |
---|---|
Name | S. aureus Peptide deformylase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | def |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16073590
|
enQuireBio™
QP6478-ye-10ug |
10 μg | N/A | ||||||
16013600
|
enQuireBio™
QP6478-ye-50ug |
50 μg | N/A | ||||||
16063590
|
enQuireBio™
QP6478-ye-100ug |
100 μg |
7930.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16093590
|
enQuireBio™
QP6478-ye-200ug |
200 μg | N/A | ||||||
16003600
|
enQuireBio™
QP6478-ye-500ug |
500 μg | N/A | ||||||
16083590
|
enQuireBio™
QP6478-ye-1mg |
1 mg |
29485.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WP_000957037.1 | |
Research Use Only | |
def | |
Recombinant Protein | |
Yeast | |
MLTMKDIIRD GHPTLRQKAA ELELPLTKEE KETLIAMREF LVNSQDEEIA KRYGLRSGVG LAAPQINISK RMIAVLIPDD GSGKSYDYML VNPKIVSHSV QEAYLPTGEG CLSVDDNVAG LVHRHNRITI KAKDIEGNDI QLRLKGYPAI VFQHEIDHLN GVMFYDHIDK NHPLQPHTDA VEV | |
Greater than 90% as determined by SDS-PAGE. |
S. aureus Peptide deformylase Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. aureus Peptide deformylase Protein