missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Gamma-hemolysin component C Protein
A DNA sequence encoding the Staphylococcus aureus (strain MRSA252) Gamma-hemolysin component C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7736-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000916697.1 | |
50 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Gamma-hemolysin component C Protein | |
Research Use Only | |
hlgC | |
Recombinant Protein | |
E. coli | |
ANDTEDIGKG NDVEIIKRTE DKTSNKWGVT QNIQFDFVKD KKYNKDALIL KMQGFISSRT TYYNYKNTNH IKSMRWPFQY NIGLKTNDKY VSLINYLPKN KIESTNVSQT LGYNIGGNFQ SAPSLGGNGS FNYSKSISYT QQNYVSEVEQ QNSKSVLWGV KANSFATESG QKSAFDSDLF VGYKPHSKDP RDYFVPDSEL PPLVQSGFNP SFIATVSHEK GSSDTSEFEI TYGRNMDVTH AIKRSTHYGN SYLDGHRVHN AFKNRNYTVK YEVNWKTHEI KVKGQN | |
Greater than 80% as determined by SDS-PAGE. |