missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus fabD Protein
A DNA sequence encoding the Staphylococcus aureus (strain COL) fabD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7663-ec-500ug
This item is not returnable.
View return policy
Specifications
WP_000047348.1 | |
500 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus fabD Protein | |
Research Use Only | |
fabD | |
Recombinant Protein | |
E. coli | |
MSKTAIIFPG QGAQKVGMAQ DLFNNNDQAT EILTSAANTL DFDILETMFT DEEGKLGETE NTQPALLTHS SALLAALKNL NPDFTMGHSL GEYSSLVAAD VLSFEDAVKI VRKRGQLMAQ AFPTGVGSMA AVLGLDFDKV DEICKSLSSD DKIIEPANIN CPGQIVVSGH KALIDELVEK GKSLGAKRVM PLAVSGPFHS SLMKVIEEDF SSYINQFEWR DAKFPVVQNV NAQGETDKEV IKSNMVKQLY SPVQFINSTE WLIDQGVDHF IEIGPGKVLS GLIKKINRDV KLTSIQTLED VKGWNEND | |
Greater than 90% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant S. aureus fabD Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction