missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus fabD Protein

Product Code. 16000061
missing translation for 'orderingAttributeHoverText'
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16000061

missing translation for 'mfr': enQuireBio™ QP7663ec200ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

A DNA sequence encoding the Staphylococcus aureus (strain COL) fabD, was expressed in the hosts and tags indicated.

Specifications

Adgangsnummer WP_000047348.1
Navn S. aureus fabD Protein
Mængde 200 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol fabD
Protein længde Full Length
Produkttype Recombinant Protein
Krydsreaktivitet S. aureus
Arter E. coli
Protein tag His-SUMO
Sekvens MSKTAIIFPG QGAQKVGMAQ DLFNNNDQAT EILTSAANTL DFDILETMFT DEEGKLGETE NTQPALLTHS SALLAALKNL NPDFTMGHSL GEYSSLVAAD VLSFEDAVKI VRKRGQLMAQ AFPTGVGSMA AVLGLDFDKV DEICKSLSSD DKIIEPANIN CPGQIVVSGH KALIDELVEK GKSLGAKRVM PLAVSGPFHS SLMKVIEEDF SSYINQFEWR DAKFPVVQNV NAQGETDKEV IKSNMVKQLY SPVQFINSTE WLIDQGVDHF IEIGPGKVLS GLIKKINRDV KLTSIQTLED VKGWNEND
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.