Learn More
enQuireBio™ Recombinant S. aureus Exfoliative toxin A Protein
A DNA sequence encoding the Staphylococcus aureus Exfoliative toxin A, was expressed in the hosts and tags indicated.
2300.00 DKK - 24645.00 DKK
Specifications
Accession Number | WP_001065781.1 |
---|---|
Name | S. aureus Exfoliative toxin A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | eta |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098090
|
enQuireBio™
QP7304-ec-10ug |
10 ÎĽg | N/A | ||||||
16038100
|
enQuireBio™
QP7304-ec-50ug |
50 ÎĽg | N/A | ||||||
16088090
|
enQuireBio™
QP7304-ec-100ug |
100 ÎĽg |
7110.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16018100
|
enQuireBio™
QP7304-ec-200ug |
200 ÎĽg | N/A | ||||||
16028100
|
enQuireBio™
QP7304-ec-500ug |
500 ÎĽg | N/A | ||||||
16008100
|
enQuireBio™
QP7304-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WP_001065781.1 | |
Research Use Only | |
eta | |
Recombinant Protein | |
E. coli | |
EVSAEEIKKH EEKWNKYYGV NAFNLPKELF SKVDEKDRQK YPYNTIGNVF VKGQTSATGV LIGKNTVLTN RHIAKFANGD PSKVSFRPSI NTDDNGNTET PYGEYEVKEI LQEPFGAGVD LALIRLKPDQ NGVSLGDKIS PAKIGTSNDL KDGDKLELIG YPFDHKVNQM HRSEIELTTL SRGLRYYGFT VPGNSGSGIF NSNGELVGIH SSKVSHLDRE HQINYGVGIG NYVKRIINEK NE | |
Greater than 80% as determined by SDS-PAGE. |
S. aureus Exfoliative toxin A Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
enQuireBio™ Recombinant S. aureus Exfoliative toxin A Protein