Learn More
enQuireBio™ Recombinant Rotavirus A Outer capsid protein VP4 Protein
A DNA sequence encoding the Rotavirus A (strain Human/United Kingdom/ST3/1975 G4-P2A[6]-I1-R1-C1-M1-A1-N1-T1-E1-H1) (RV-A) (Rotavirus A (strain St. Thomas 3)) Rotavirus A Outer capsid protein VP4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7052-ec-200ug
Specifications
Rotavirus A Outer capsid protein VP4 Protein | |
Research Use Only | |
Full Length | |
Rotavirus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
200 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
AQVSEDIIIS KTSLWKEMQY NRDIIIRFKF NNSIIKLGGL GYKWSEISFK AANYQYNYLR DGEQVTAHTT CSVNGVNNFS YNGGLLPTHF SISRYEVIKE NSYVYVDYWD DSQAFRNMVY VRSLAANLNS VKCSGGNYNF QMPVGAWPVM SGGAVSLHFA GVTLSTQFTD FVSLNSLRFR FSLTVEEPPF SILRTRVSGL YGLPASNPNS GHEYYEIAGR FSLISLVPSN DDY | |
Greater than 90% as determined by SDS-PAGE. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.