Learn More
enQuireBio™ Recombinant Rotavirus A Intermed. capsid protein VP6 Protein
A DNA sequence encoding the Rotavirus A (strain Human/United Kingdom/ST3/1975 G4-P2A[6]-I1-R1-C1-M1-A1-N1-T1-E1-H1) (RV-A) (Rotavirus A (strain St. Thomas 3)) Rotavirus A Intermed. capsid protein VP6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7509-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
Rotavirus A Intermed. capsid protein VP6 Protein | |
Research Use Only | |
Full Length | |
Rotavirus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
200 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
MEVLYSLSKT LKDARDKIVE GTLYSNVSDL IQQFNQMIVT MNGNDFQTGG IGNLPIRNWT FDFGLLGTTL LNLDANYVET ARTTIEYFID FIDNVCMDEM ARESQRNGVA PQSEALRKLA GIKFKRINFN NSSEYIENWN LQNRRQRTGF VFHKPNIFPY SASFTLNRSQ PMHDNLMGTM WLNAGSEIQV AGFDYSCALN APANIQQFEH IVQLRRALTT ATITLLPDAE RFSFPRVINS ADGATTWFFN PIILRPNNVE VEFLLNGQII NTYQARFGTI IARNFDTIRL SFQLMRPPNM TPAVNALFPQ AQPFQHHATV GLTLRIESAV CESVLADANE TLLANVTAVR QEYAIPVGPV FPPGMNWTEL ITNYSPSRED NLQRVFTVAS IRSMLIK | |
Greater than 90% as determined by SDS-PAGE. |