missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Vesicle-fusing ATPase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Vesicle-fusing ATPase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8832-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_068516.1 | |
Vesicle-fusing ATPase Protein | |
Research Use Only | |
Nsf | |
Recombinant Protein | |
E. coli | |
QAARCPTDEL SLSNCAVVNE KDYQSGQHVM VRTSPNHKYI FTLRTHPSVV PGCIAFSLPQ RKWAGLSIGQ DIEVALYSFD KAKQCIGTMT IEIDFLQKKN IDSNPYDTDK MAAEFIQQFN HQAFSVGQQL VFSFNDKLFG LLVKDIEAMD PSILKGEPAS GKRQKIEVGL VVGNSQVAFE KAENSSLNLI GKAKTKENRQ SII | |
Greater than 90% as determined by SDS-PAGE. |
60355 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rat Vesicle-fusing ATPase Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction