Få mere at vide
enQuireBio™ Recombinant Rat Vasopressin V1a receptor Protein Artikelnummer.: 16076781
enQuireBio™ Recombinant Rat Vasopressin V1a receptor Protein
Artikelnummer. 16076781
500 μg, 500µg
Mængde:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
10µg
1mg
200µg
500µg
50µg
Artikelnummer. 16076781
Brand: enQuireBio™ QP8843ec500ug
Antal Pris
1 12730.00 DKK / 500µg
3 11902.55 DKK / 500µg
Se alternative produkter
A DNA sequence encoding the Rattus norvegicus (Rat) Vasopressin V1a receptor, was expressed in the hosts and tags indicated.
Tekniske data
Adgangsnummer NP_444178.2
Gen-id (Entrez) 25107
Navn Vasopressin V1a receptor Protein
Mængde 500 μg
Regulatorisk status Research Use Only
Endotoksin koncentration Not determined.
Gen symbol Avpr1a
Protein længde Partial (see sequence information for more details).
Produkttype Recombinant Protein
Krydsreaktivitet Rat
Arter E. coli
Protein tag GST
Sekvens SQDRSVGNSS PWWPLTTEGS NGSQEAARLG EGDSPLGDVR NEELAK
Buffer Tris-based buffer, 50% glycerol
Renhed eller kvalitetsklasse Greater than 90% as determined by SDS-PAGE.
Produkttitel
enQuireBio™ Recombinant Rat Vasopressin V1a receptor Protein >