missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat REG3B / Pap1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) REG3B / Pap1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7120-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087430

  • 3370.00 DKK / 50µg
Estimated Shipment: 24-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_445741.1
REG3B / Pap1 Protein
50 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
24618
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Reg3b
Recombinant Protein
E. coli
EDSPKKIPSA RISCPKGSQA YGSYCYALFQ IPQTWFDAEL ACQKRPEGHL VSVLNVAEAS FLASMVKNTG NSYQYTWIGL HDPTLGGEPN GGGWEWSNND IMNYVNWERN PSTALDRGFC GSLSRSSGFL RWRDTTCEVK LPYVCKFTG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers