missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PROS1 / Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PROS1 /, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6534-ma-20ug
Additional Details : Weight : 0.01000kg
Specifications
PROS1 / Protein | |
20 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
PROS1 | |
Recombinant Protein | |
Mammal | |
ANTLLEETKK GNLERECIEE LCNKEEAREV FENNPETDYF YPKYLGCLGA FRVGAFSAAR QSANAYPDLR SCVNAIPDQC DPMPCNEDGY LSCKDGQGAF TCICKPGWQG DKCQFDINEC KDPSNINGGC SQTCDNTPGS YHCSCKIGFA MLTNKKDCKD VDECSLKPSV CGTAVCKNIP GDFECECPNG YRYDPSSKSC KDVDECSENT CAQLCVNYPG GYSCYCDGKK GFKLAQDQRS CEGIPVCLSL DLDKNYELLY LAEQFAGVVL YLKFRLPDIT RFSAEFDFRT YDSEGIILYA ESLDHSNWLL IALREGKIEV QFKNEFSTQI TTGGNVINNG IWNMVSVEEL DDSVSIKIAK EAVMNINKLG SLFKPTDGFL DTKIYFAGLP RKVESALIKP INPRLDGCIR GWNLMKQGAL GAKEIVEGKQ NKHCFLTVEK GSYYPGSGIA QFSIDYNNVT NAEGWQINVT LNIRPSTGTG VMLALVSGDT VPFALSLVDS GSGTSQDILV FVENSVAAHL EAITLCSEQP SQLKCNINRN GLELWTPVRK DVIYSKDLQR QLAILDKTMK GTVATYLGGV PDISFSATPV NAFYSGCMEV NINGVQLDLD EAISKHNDIR AHSCPSVRKI QKNF |