missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PLAUR / CD87 / uPAR Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PLAUR / CD87 / uPAR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6504-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_059046.1 | |
PLAUR / CD87 / uPAR Protein | |
Research Use Only | |
PLAUR | |
Recombinant Protein | |
E. coli | |
LRCIQCESNQ DCLVEECALG QDLCRTTVLR EWEDAEELEV VTRGCAHKEK TNRTMSYRMG SVIVSLTETV CATNLCNRPR PGARGRPFPR GRYLECASCT SLDQSCERGR EQSLQCRYPT EHCIEVVTLQ STERSVKDEP YTKGCGSLPG CPGTAGFHSN QTFHFLKCCN FTQCNGGP | |
Greater than 90% as determined by SDS-PAGE. |
50692 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |