missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PLA2G2A Protein

A DNA sequence encoding the Rattus norvegicus (Rat) PLA2G2A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7058-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_113786.3
PLA2G2A Protein
Research Use Only
Pla2g2a
Recombinant Protein
E. coli
SLLEFGQMIL FKTGKRADVS YGFYGCHCGV GGRGSPKDAT DWCCVTHDCC YNRLEKRGCG TKFLTYKFSY RGGQISCSTN QDSCRKQLCQ CDKAAAECFA RNKKSYSLKY QFYPNKFCKG KTPSC
Greater than 90% as determined by SDS-PAGE.
29692
10 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers