missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat p53 / TP53 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) p53 / TP53, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8835-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_112251.2 | |
p53 / TP53 Protein | |
Research Use Only | |
TP53 | |
Recombinant Protein | |
E. coli | |
MEDSQSDMSI ELPLSQETFS CLWKLLPPDD ILPTTATGSP NSMEDLFLPQ DVAELLEGPE EALQVSAPAA QEPGTEAPAP VAPASATPWP LSSSVPSQKT YQGNYGFHLG FLQSGTAKSV MCTYSISLNK LFCQLAKTCP VQLWVTSTPP PGTRVRAMAI YKKSQHMTEV VRRCPHHERC SDGDGLAPPQ HLIRVEGNPY AEYLDDRQTF RHSVVVPYEP PEVGSDYTTI HYKYMCNSSC MGGMNRRPIL TIITLEDSSG NLLGRDSFEV RVCACPGRDR RTEEENFRKK EEHCPELPPG SAKRALPTST SSSPQQKKKP LDGEYFTLKI RGRERFEMFR ELNEALELKD ARAAEESGDS RAHSSYPKTK KGQSTSRHKK PMIKKVGPDS D | |
Greater than 90% as determined by SDS-PAGE. |
24842 | |
200 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rat p53 / TP53 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction