missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Lumican / LUM Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Lumican / LUM, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6324-ma-50ug
This item is not returnable.
View return policy
Specifications
NP_112312.1 | |
Lumican / LUM Protein | |
Research Use Only | |
Lum | |
Recombinant Protein | |
Mammal | |
QYYDYDAPLF MYGELSPNCA PECNCPHSYP TAMYCDDLKL KSVPMVPPGI KYLYLRNNQI DHIDEKAFEN VTDLQWLILD HNLLENSKIK GKVFSKLKQL KKLHINYNNL TESVGPLPKS LQDLQLANNK ISKLGSFDGL VNLTFIYLQH NQLKEEAVSA SLKGLKSLEY LDLSFNQMSK LPAGLPTSLL TLYLDNNKIT NIPDEYFNRF TGLQYLRLSH NELADSGVPG NSFNISSLLE LDLSYNKLKS IPTVNENLEN YYLEVNKLEK FDVKSFCKIL GPLSYSKIKH LRLDGNPLTQ SSLPPDMYEC LRVANEITVN | |
Greater than 90% as determined by SDS-PAGE. |
81682 | |
50 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Rat Lumican / LUM Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction