missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Galectin-4 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Galectin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6297-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16092260

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_037107.1
Galectin-4 Protein
Research Use Only
Lgals4
Recombinant Protein
E. coli
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSIYIQGIAK DNMRRFHVNF AVGQDEGADI AFHFNPRFDG WDKVVFNTMQ SGQWGKEEKK KSMPFQKGHH FELVFMVMSE HYKVVVNGTP FYEYGHRLPL QMVTHLQVDG DLELQSINFL GGQPAASQYP GTMTIPAYPS AGYNPPQMNS LPVMAGPPIF NPPVPYVGTL QGGLTARRTI IIKGYVLPTA KNLIINFKVG STGDIAFHMN PRIGDCVVRN SYMNGSWGSE ERKIPYNPFG AGQFFDLSIR CGTDRFKVFA NGQHLFDFSH RFQAFQRVDM LEIKGDITLS YVQI
Greater than 90% as determined by SDS-PAGE.
25474
10 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers