missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Galectin-4 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Galectin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6297-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037107.1 | |
Galectin-4 Protein | |
Research Use Only | |
Lgals4 | |
Recombinant Protein | |
E. coli | |
MAYVPAPGYQ PTYNPTLPYK RPIPGGLSVG MSIYIQGIAK DNMRRFHVNF AVGQDEGADI AFHFNPRFDG WDKVVFNTMQ SGQWGKEEKK KSMPFQKGHH FELVFMVMSE HYKVVVNGTP FYEYGHRLPL QMVTHLQVDG DLELQSINFL GGQPAASQYP GTMTIPAYPS AGYNPPQMNS LPVMAGPPIF NPPVPYVGTL QGGLTARRTI IIKGYVLPTA KNLIINFKVG STGDIAFHMN PRIGDCVVRN SYMNGSWGSE ERKIPYNPFG AGQFFDLSIR CGTDRFKVFA NGQHLFDFSH RFQAFQRVDM LEIKGDITLS YVQI | |
Greater than 90% as determined by SDS-PAGE. |
25474 | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |