missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Rat Fetuin-B / FETUB Protein Product Code.: 16098951

enQuireBio™ Recombinant Rat Fetuin-B / FETUB Protein

Product Code. 16098951
50 μg, 50µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16098951

Brand: enQuireBio™ QP9897ye50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Rattus norvegicus (Rat) Fetuin-B / FETUB, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_445800.2
Gene ID (Entrez) 83928
Name Fetuin-B / FETUB Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Fetub
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Rat
Species Yeast
Protein Tag His
Sequence RSPPAPPLPN APFAPLRPLG CNDSEVLAVA GFALQNINRV QKDGYMLTLN RVHDARVHRQ EDMGSLFYLM LDVLETGCHV LSRKALKDCG PRIFYETVHG QCKAMFHVNK PRRVLYLPAY NCTLRPVSKR KIHSMCPDCP HPVDLSAPSV LEAATESLAK FNSENPSKQY ALVKVTKATT QWVVGPSYFV EYLIKESPCT QSQDSCSLQA SDSEPVGLCQ GSLIKSPGVP PQRFKKTVTV SCEFFESQDQ VPGGENPADT QDAKKLPQKN TAPTSSPSIT APRGSIQHLP EQEEPEDSKG KSPEEPFPVQ LDLTTNPQGD TLDVSFLYLE PEEKKLVVLP FPGKEQRSPE CPGPEKQRTP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat Fetuin-B / FETUB Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.